missing translation for 'onlineSavingsMsg'
Learn More

(-)-Limonene, 96%

Product Code. p-3758280 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10402211

Brand: Thermo Scientific Acros 203731000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5989-54-8 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5989-54-8
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001558
InChI Key XMGQYMWWDOXHJM-SNVBAGLBSA-N
Synonym --limonene, l-limonene, s---limonene, s-limonene, --4s-limonene, --s-limonene, s-p-mentha-1,8-diene, unii-47maj1y2ne, 4s-limonene, p-mentha-1,8-diene, s--
PubChem CID 439250
ChEBI CHEBI:15383
IUPAC Name (4S)-1-methyl-4-prop-1-en-2-ylcyclohexene
SMILES CC1=CCC(CC1)C(=C)C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point -104°C to 84°C
Color Colorless to Yellow
Density 0.8420g/mL
Boiling Point 175.0°C to 177.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4700 to 1.4740
Quantity 100 mL
Beilstein 05, 136
Merck Index 14, 5493
Specific Gravity 0.842
Specific Rotation Condition - 99.50 (20.00°C c=neat)
Specific Rotation − 99.50
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with alcohol
Formula Weight 136.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material (-)-Limonene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (-)-Limonene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : L-Limonene, D-Limonene, p-Cymene, 1,8-Cineol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.