missing translation for 'onlineSavingsMsg'
Learn More

LIMKi 3, TRC

Product Code. 30475729
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 30475729

Brand: Toronto Research Chemicals TRCL3976881MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[5-[2-(2,6-dichlorophenyl)-5-(difluoromethyl)pyrazol-3-yl]-1,3-thiazol-2-yl]-2-methylpropanamide
CAS 1338247-35-0
Molecular Formula C17 H14 Cl2 F2 N4 O S
Quantity 1 mg
Synonym LIMKi 3, N-[5-[1-(2,6-Dichlorophenyl)-3-(difluoromethyl)-1H-pyrazol-5-yl]-2-thiazolyl]-2-methylpropanamide, BMS 5
InChI Formula InChI=1S/C17H14Cl2F2N4OS/c1-8(2)16(26)23-17-22-7-13(27-17)12-6-11(15(20)21)24-25(12)14-9(18)4-3-5-10(14)19/h3-8,15H,1-2H3,(H,22,23,26)
SMILES CC(C)C(=O)Nc1ncc(s1)c2cc(nn2c3c(Cl)cccc3Cl)C(F)F
IUPAC Name N-[5-[2-(2,6-dichlorophenyl)-5-(difluoromethyl)pyrazol-3-yl]-1,3-thiazol-2-yl]-2-methylpropanamide
Molecular Weight (g/mol) 431.287
Formula Weight 430.023
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.