missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIMKi 3, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | N-[5-[2-(2,6-dichlorophenyl)-5-(difluoromethyl)pyrazol-3-yl]-1,3-thiazol-2-yl]-2-methylpropanamide |
| CAS | 1338247-35-0 |
| Molecular Formula | C17 H14 Cl2 F2 N4 O S |
| Quantity | 10 mg |
| Synonym | LIMKi 3, N-[5-[1-(2,6-Dichlorophenyl)-3-(difluoromethyl)-1H-pyrazol-5-yl]-2-thiazolyl]-2-methylpropanamide, BMS 5 |
| InChI Formula | InChI=1S/C17H14Cl2F2N4OS/c1-8(2)16(26)23-17-22-7-13(27-17)12-6-11(15(20)21)24-25(12)14-9(18)4-3-5-10(14)19/h3-8,15H,1-2H3,(H,22,23,26) |
| SMILES | CC(C)C(=O)Nc1ncc(s1)c2cc(nn2c3c(Cl)cccc3Cl)C(F)F |
| IUPAC Name | N-[5-[2-(2,6-dichlorophenyl)-5-(difluoromethyl)pyrazol-3-yl]-1,3-thiazol-2-yl]-2-methylpropanamide |
| Molecular Weight (g/mol) | 431.287 |
| Formula Weight | 430.023 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction