missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIMK2 Recombinant Human Protein
LIMK2 Recombinant Human Protein
Brand: Invitrogen PV3860
Code : 50
Additional Details : Weight : 0.00900kg
Description
LIMK2 is a serine⁄threonine kinase involved in regulation of actin cytoskeletal changes via phosphorylation of cofilin. LIMK2 has also been implicated in brain development.Specifications
NP_005560 | |
Kinase Assay, Protein Assays and Analysis, Target and Lead Identification and Validation | |
LIMK2 Recombinant Human Protein | |
–80°C | |
1 Each | |
LIMK2 Recombinant Human Protein | |
LISK | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.14mg/mL | |
99.7kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
LIMK2 | |
The ability of LIMK2 to bind the ATP competitor kinase tracer 178 (PV5593) was demonstrated for this lot of LIMK2 in the LanthaScreen™ Eu kinase binding assay. Increasing amounts of kinase tracer 178 (PV5593) were incubated with 5nM LIMK2 and 2nM Eu-anti-GST antibody (PV5594) in kinase buffer A (PV3189) with and without the presence of 10μM dasatinib as the competitor for the kinase tracer | |
LIMK2 Recombinant Human Protein | |
GST Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.