missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIMK1 Recombinant Human Protein
LIMK1 Recombinant Human Protein
Brand: Invitrogen PV4337
Code : 50
Additional Details : Weight : 0.00945kg
Description
LIMK1 is a serine⁄threonine kinase involved in regulation of actin cytoskeletal changes via phosphorylation of cofilin. LIMK1 has also been implicated in brain development.Specifications
NP_002305 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
44.7kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
LIMK1, LIMK | |
LIMK1 was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL Cofilin 2 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 285-638 | |
98nmoles of phosphate transferred to Cofilin 2 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2.03μg/mL | |
His Tag | |
Kinase and inhibitors | |
70% by SDS-PAGE |
0.38mg/mL | |
3984 | |
LIMK1 Recombinant Human Protein | |
–80°C | |
1 Each | |
LIMK1 Recombinant Human Protein | |
LISK | |
LIMK1 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
For Research Use Only. Not for use in diagnostic procedures.