missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIMK1 Monoclonal Antibody (OTI6B4)

LIMK1 Monoclonal Antibody (OTI6B4)
Brand: Invitrogen MA525569
This item is not returnable.
View return policy
Description
LIMK1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
| LIMK1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P53667, P53668, P53669 | |
| LIMK1 | |
| Full length human recombinant protein of LIMK1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI6B4 | |
| Unconjugated | |
| LIMK1 | |
| EC 2.7.11.1; kinase LIMK1; KIZ-1; LIM domain kinase 1; lim kinase 1; LIM motif-containing protein kinase; LIM motif-containing protein kinase 1; LIM-domain containing protein kinase 1; LIM-domain containing, protein kinase; LIM-domain containing, protein kinase 1; Limk; LIMK1; LIMK-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 16885, 3984, 489800, 65172 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction