missing translation for 'onlineSavingsMsg'
Learn More

Limitin/IFN-zeta Antibody (RM0123-7D19), PE, Novus Biologicals™

Product Code. 18432632 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432632 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432632 Supplier Novus Biologicals Supplier No. NBP212426PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Limitin/IFN-zeta Monoclonal antibody specifically detects Limitin/IFN-zeta in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen Limitin/IFN-zeta
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
Classification Monoclonal
Clone RM0123-7D19
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize
Formulation PBS
Gene Alias 6030405N23Rik, BGIF, Ifnz, interferon zeta, Lmtn
Host Species Rat
Immunogen Mouse recombinant Limitin/IFN-zeta
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319146
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.