missing translation for 'onlineSavingsMsg'
Learn More

LIME Antibody (mLIME-05), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18417235 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417235 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417235 Supplier Novus Biologicals Supplier No. NBP147230JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LIME Monoclonal antibody specifically detects LIME in Mouse samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIME
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone mLIME-05
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias dJ583P15.4, FLJ20406, Lck interacting transmembrane adaptor 1, Lck-interacting membrane protein, Lck-interacting molecule, lck-interacting transmembrane adapter 1, LIMELP8067
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 86-202 of mouse LIME
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54923
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.