missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIMD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-80518
This item is not returnable.
View return policy
Description
LIMD2 Polyclonal specifically detects LIMD2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LIMD2 | |
| Polyclonal | |
| Western Blot 1:1000, Immunohistochemistry 1:1000, Immunohistochemistry-Paraffin 1:1000 | |
| LIM domain containing 2, LIM domain-containing protein 2, MGC10986 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| 80774 | |
| Human, Mouse, Rat |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Lyophilized from whole serum | |
| LIMD2 | |
| A synthetic peptide from human LIMD2 conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat and mouse. | |
| 0.1 mL | |
| Primary | |
| Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction