missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIMD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28106-1-AP-150UL
This item is not returnable.
View return policy
Description
Proteintech's 28106-1-AP antibody was generated by 133 amino acids, it could recognize the full-length protein in applications.
The Zyxin family of proteins contains five members: Ajuba, Limd1, LPP, TRIP6 and Zyxin. Limd1 (LIM domain-containing protein 1) is a ubiquitously expressed tumor suppressor containing three LIM zinc-binding domains. LIM domains consist of a cysteine-rich consensus sequence containing two distinct zinc-binding subdomains, which mediate protein-protein interactions. Limd1 interacts with the proteins SQSTM1, Rb, p62 and TRAF6. Limd1 was first identified when the deletion of its gene was noted in some cervical cancers. Limd1 blocks in vitro and in vivo tumor growth and is downregulated in lung cancer.Specifications
| LIMD1 | |
| Polyclonal | |
| Unconjugated | |
| LIMD1 | |
| LIM domains containing 1, LIMD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8994 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UGP4 | |
| LIMD1 | |
| LIMD1 Fusion Protein Ag27974 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction