missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586896
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
This gene encodes a member of a large protein family which contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in control of differentiation and development of neural and lymphoid cells. A similar protein in mice is an essential regulator of the vertebrate head organizer.
Specifications
| LIM1 | |
| Polyclonal | |
| Unconjugated | |
| Lhx1 | |
| hLim-1; Homeobox protein Lim-1; LHX1; LHX-1; LIM homeo box protein 1; LIM homeobox 1; LIM homeobox protein 1; LIM/homeobox protein Lhx1; Lim1; Lim-1; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx1; MGC126723; MGC138141; rlim; transcription factor; homeobox protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16869, 257634, 3975 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P48742, P63006, P63007 | |
| Lhx1 | |
| A synthetic peptide corresponding to residues in Human LHX1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction