missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546855
This item is not returnable.
View return policy
Description
Peptide sequence: PEPSLPGPLH SMSAEVFGPS PPFSSLSVNG GASYGNHLSH PPEMNEAAVW Sequence homology: Cow: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 85%.
This gene encodes a member of a large protein family which contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and be involved in control of differentiation and development of neural and lymphoid cells. A similar protein in mice is an essential regulator of the vertebrate head organizer.
Specifications
| LIM1 | |
| Polyclonal | |
| Unconjugated | |
| Lhx1 | |
| hLim-1; Homeobox protein Lim-1; LHX1; LHX-1; LIM homeo box protein 1; LIM homeobox 1; LIM homeobox protein 1; LIM/homeobox protein Lhx1; Lim1; Lim-1; LOW QUALITY PROTEIN: LIM/homeobox protein Lhx1; MGC126723; MGC138141; rlim; transcription factor; homeobox protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3975 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P48742 | |
| Lhx1 | |
| Synthetic peptide directed towards the C-terminal of human LHX1 (aa 357-406). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction