missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£356.00 - £559.00
Specifications
| Antigen | LIM1 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619654
|
Novus Biologicals
NBP3-21271-100ul |
100 μg |
£559.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18612815
|
Novus Biologicals
NBP3-21271-25ul |
25 μg |
£356.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIM1 Polyclonal antibody specifically detects LIM1 in Human samples. It is validated for ImmunofluorescenceSpecifications
| LIM1 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, DNA replication Transcription Translation and Splicing, Neuroscience | |
| PBS, pH 7.2, 40% glycerol | |
| 3975 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| hLim-1, Homeobox protein Lim-1, LIM homeobox 1, LIM homeobox protein 1, LIM-1LIM/homeobox protein Lhx1, LIM1MGC138141, MGC126723 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: NGPFSFYGDYQSEYYGPGGNYDFFPQGPPSSQAQTPVDLPFVPSSGPSGTPLGGLEHPLPGHHPSSEAQRFTDILAHPPGDSPSPEPSLPGPLHSMSAEVFGPSPPFSSLSVNGGASYGNHL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title