missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM Kinase 1 Antibody (SR2175), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21902-100ul
This item is not returnable.
View return policy
Description
LIM Kinase 1 Monoclonal antibody specifically detects LIM Kinase 1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| LIM Kinase 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| SR2175 | |
| Western Blot 1:500-1:2000 | |
| EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1 | |
| A synthesized peptide derived from human LIM Kinase 1 (Uniprot #: P53667) | |
| 100 μg | |
| Breast Cancer, Cancer, Phospho Specific, Protein Kinase | |
| 3984 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction