missing translation for 'onlineSavingsMsg'
Learn More

LIM domain only 3 Antibody, Novus Biologicals™

Product Code. 18344060 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344060 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344060 Supplier Novus Biologicals Supplier No. NBP275090

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

LIM domain only 3 Polyclonal specifically detects LIM domain only 3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIM domain only 3
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA 1:128000, Immunofluorescence 10 ug/mL
Formulation TBS (pH 7.3), 0.5% BSA with 0.02% Sodium Azide
Gene Alias DAT1, LIM domain only 3 (rhombotin-like 2), LMO-3, neuronal specific transcription factor DAT1, Neuronal-specific transcription factor DAT1, RBTN3, RBTNL2MGC26081, Rhom-3, RHOM3LIM domain only protein 3, rhombotin-3, rhombotin-like 2
Gene Symbols LMO3
Host Species Goat
Immunogen Peptide with sequence C-EGLMKEGYAPQVR, from the C Terminus of the protein sequence according to NP_997105.1.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 55885.0
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.