Learn More
LILRB5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552906
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PKPTLWAEPA SVIARGKPVT LWCQGPLETE EYRLDKEGLP WARKRQNPLE PGAKAKFHIP STVYDSAGRY RCYYETPAGW SEPSDPLELV ATG Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 56%.
LILRB5 Polyclonal antibody specifically detects LILRB5 in Human samples. It is validated for Immunohistochemistry.Specifications
LILRB5 | |
Polyclonal | |
Unconjugated | |
LILRB5 | |
CD85 antigen-like family member C; CD85C; leucocyte Ig-like receptor B5; leukocyte immunoglobulin like receptor B5; leukocyte immunoglobulin-like receptor 8; Leukocyte immunoglobulin-like receptor subfamily B member 5; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 5; LILRB5; LIR8; LIR-8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10990 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75023 | |
LILRB5 | |
Recombinant protein corresponding to Human LILRB5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only