missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LILRB4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598762
This item is not returnable.
View return policy
Description
LILRB4 Polyclonal Antibody for Western Blot, ELISA
This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. The receptor can also function in antigen capture and presentation. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].
Specifications
| LILRB4 | |
| Polyclonal | |
| Unconjugated | |
| LILRB4 | |
| CD85 antigen-like family member K; CD85K; glycoprotein 49 B; glycoprotein 49b; gp49; Gp49b; GP49B2; HM18; ILT3; ILT-3; Immunoglobulin-like transcript 3; leucocyte Ig-like receptor B4; leukocyte immunoglobulin like receptor B4; leukocyte immunoglobulin-like receptor 5; Leukocyte immunoglobulin-like receptor subfamily B member 4; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 4; leukocyte immunoglobulin-like receptor, subfamily B, member 4; leukocyte surface receptor; Lilrb4; LIR5; LIR-5; mast cell surface glycoprotein Gp49B; Monocyte inhibitory receptor HM18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11006 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NHJ6 | |
| LILRB4 | |
| Recombinant Human Leukocyte immunoglobulin-like receptor subfamily B member 4 protein (22-259AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction