missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LILRB3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5144736

Product Code. 18005621

  • £522.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: CHYYSSAGWS EPSDPLELVM TGFYNKPTLS ALPSPVVASG GNMTLRCGSQ KGYHHFVLMK EGEHQLPRTL DSQQLHSGGF QALFPVGPVT PSHRWRFTCY YYYTNTPWVW SHPSDPLEIL PS Antigen sequence identity: Mouse 55%; Rat 57%.

This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LILRB3
Polyclonal
Unconjugated
LILRB3
CD85 antigen-like family member A; CD85A; Cell-surface glycoprotein p91; Gp91; HL9; ILT5; ILT-5; immunoglobulin-like transcript 5; leucocyte Ig-like receptor B3; leukocyte immunoglobulin like receptor B3; leukocyte immunoglobulin-like receptor 3; leukocyte immunoglobulin-like receptor subfamily B member 3; leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 6; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3; LILRA6; Lilrb3; LIR3; LIR-3; monocyte inhibitory receptor HL9; paired Ig-like receptor B; paired immunoglobulin-like receptor B; PIRB; PIR-B
Rabbit
Affinity chromatography
RUO
11025
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
IgG
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O75022
LILRB3
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 98-219. Recombinant protein control fragment (Product #RP-110177).
100 μL
Primary
Human
Antibody
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.