missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LILRA4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21153-1-AP-20UL
This item is not returnable.
View return policy
Description
ILT7 is a pDC-specific IgG-like transcript and is a member of the leukocyte immunoglobulin-like receptor gene family. This receptor, in complex with FcER1a, transduces ITAM-mediated signals which negatively modulate type I IFN production by pDC. ILT7 is thus a potential target to regulate Type 1 IFN production by pDC, which play a major role during viral infection, auto-immune diseases and anti-cancer immunity. Additionally, ILT7, along with CD123 and BDCA-2, can be used to identify or classify subsets of pDCalong with TLR expression and TLR mediated immune responses.Specifications
| LILRA4 | |
| Polyclonal | |
| Unconjugated | |
| LILRA4 | |
| CD85g, ILT 7, ILT7, LILRA4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23547 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P59901 | |
| LILRA4 | |
| LILRA4 Fusion Protein Ag15721 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction