missing translation for 'onlineSavingsMsg'
Learn More

Lilolidine, 98%, Thermo Scientific™

Product Code. 15592313 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15592313 5 g 5g
15582313 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15592313 Supplier Thermo Scientific Acros Supplier No. 452900050
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 5840-01-7
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.21
InChI Key QCCKSFHMARIKSK-UHFFFAOYSA-N

Specifications

Loss on Drying 1% max. (60°C) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Synonym lilolidine, 5,6-dihydro-4h-pyrrolo 3,2,1-ij quinoline, 1,7-trimethyleneindole, 2,3-dihydro-1h-pyrrolo 3,2,1-ij quinoline, 4h-pyrrolo 3,2,1-ij quinoline, 5,6-dihydro, lilolidene, 1-azatricyclo 6.3.1.0?, 1 2 dodeca-2,4,6,8 12-tetraene, g00001-watson-int, ksc917k8l
SMILES C1CC2=CC=CC3=C2N(C1)C=C3
PubChem CID 15645629
Formula Weight 157.21
Percent Purity 98%
Chemical Name or Material Lilolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.