missing translation for 'onlineSavingsMsg'
Learn More

Lilolidine, 98%, Thermo Scientific™

Product Code. 15582313 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15582313 1 g 1g
15592313 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15582313 Supplier Thermo Scientific Acros Supplier No. 452900010
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 5840-01-7
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.21
InChI Key QCCKSFHMARIKSK-UHFFFAOYSA-N
IUPAC Name 2-methoxy-4-N-phenylbenzene-1,4-diamine

Specifications

Loss on Drying 1% max. (60°C) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Synonym 3-methoxy-n1-phenylbenzene-1,4-diamine, 2-methoxy-n4-phenyl-1,4-phenylenediamine, 4-amino-3-methoxydiphenylamine, 3-methoxy-1-n-phenylbenzene-1,4-diamine, acmc-1apzp, 4-anilino-2-methoxyaniline, p-methoxy-p-aminodiphenylamine, 1,4-benzenediamine, 2-methoxy-n4-phenyl, 2-methoxy-n∼4∼-phenylbenzene-1,4-diamine, 2-methoxy-n4-phenyl-1,4-phenylene diamine
SMILES C1CN2C=CC3=CC=CC(C1)=C23
PubChem CID 611802
Formula Weight 157.21
Percent Purity 98%
Chemical Name or Material Lilolidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.