missing translation for 'onlineSavingsMsg'
Learn More

Lignin, MedChemExpress

Product Code. 16471159
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471159 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471159 Supplier Medchem Express Supplier No. HY111830
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Lignin (Lignine) is a natural complex biopolymer with biodegradable and biocompatible. Lignin is the main component of plant cell walls and is a renewable aromatic polymer. Lignin has strongly antioxidant activity.

Lignin (Lignine) is a natural complex biopolymer with biodegradable and biocompatible. Lignin is the main component of plant cell walls and is a renewable aromatic polymer. Lignin has strongly antioxidant activity.

Specifications

Chemical Name or Material Lignin
Color Coffee
CAS 9005-53-2
Physical Form Solid
Purity Grade Notes Research
Synonym Lignine
Solubility Information DMSO : ≥ 50 mg/mL ∣H2O : < 0.1 mg/mL (insoluble)
Quantity 1 g
SMILES [Lignin]
Grade Research

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.