missing translation for 'onlineSavingsMsg'
Learn More

Lignan P, TRC

Product Code. 30134874
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134874 500 mg 500mg
30006939 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134874 Supplier Toronto Research Chemicals Supplier No. TRCL397860500MG
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23363-35-1
Molecular Formula C27 H30 O13
Molecular Weight (g/mol) 562.52
Synonym Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 9-(β-D-glucopyranosyloxy)-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-, (5R,5aR,8aR,9S)-, Epipodophyllotoxin, 4'-demethyl-, 9-β-D-glucopyranoside (8CI), Furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, 9-(β-D-glucopyranosyloxy)-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-, [5R-(5α,5aβ,8aα,9β)]-, (5R,5aR,8aR,9S)-9-(β-D-Glucopyranosyloxy)-5,8,8a,9-tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)furo[3',4':6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one, Glucopyranoside, 4'-demethylepipodophyllotoxin, β-D- (8CI), 4-Demethylepipodophyllotoxin 7'-O-β-D-glucopyranoside, 4'-Demethylepipodophyllotoxin glucoside, 4'-Demethylepipodophyllotoxin β-D-glucopyranoside, Lignan P, Pptoxin II, 4'-Demethylepipodophyllotoxin 9-(β-D-glucopyranoside), (5R,5aR,8aR,9S)-9-(β-D-Glucopyranosyloxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,8,8a,9-tetrahydro[2]benzofuro-[5,6-f][1,3]benzodioxol-6(5aH)-one
IUPAC Name (5S,5aR,8aR,9R)-9-(4-hydroxy-3,5-dimethoxyphenyl)-5-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-5a,6,8a,9-tetrahydro-5H-[2]benzofuro[5,6-f][1,3]benzodioxol-8-one
SMILES COc1cc(cc(OC)c1O)[C@H]2[C@@H]3[C@H](COC3=O)[C@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c5cc6OCOc6cc25

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C27H30O13/c1-34-16-3-10(4-17(35-2)21(16)29)19-11-5-14-15(38-9-37-14)6-12(11)25(13-8-36-26(33)20(13)19)40-27-24(32)23(31)22(30)18(7-28)39-27/h3-6,13,18-20,22-25,27-32H,7-9H2,1-2H3/t13-,18+,19+,20-,22+,23-,24+,25+,27-/m0/s1
Formula Weight 562.1686
Percent Purity >95%
Chemical Name or Material Lignan P
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.