missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid Texas Red Antibody Labeling Kit

Product Code. 18032756 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18032756

Brand: Novus Biologicals™ 3150005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Novus Biologicals™ Lightning-Link Rapid Texas Red Antibody Labeling Kit enables direct labeling of proteins, peptides or other biomolecules for use in research and development applications, drug discovery and the development of diagnostic kits.

  • The easy-to-use, one step procedure allows researchers to covalently label biomolecules with only 30 seconds hands-on time.
  • Conjugates are ready to use in less than twenty minutes.
  • Simply pipettes the biomolecule into a vial of lyophilized mixture containing the label of interest and incubates (for more details please watch the video below)
  • Texas red is a red fluorescent dye, used in histology, in fluorescence microscopy applications and in immunohistochemistry
  • It has a strong absorption at 596nm, with a high fluorescence at 616nm (extinction coefficient 8.0 x 104cm-1M-1) and high quantum yield
  • By circumventing the desalting or dialysis steps that commonly interrupt traditional antibody conjugation procedures, LightningLink technology can be used to label both small and large quantities of primary antibodies with ease
  • Batch-to-batch variation upon scale up is minimal as the process is so simple, and recoveries are always 100%
  • Includes: 1 vial of LL Rapid Modifier reagent, 1 vial of LL Rapid Quencher reagent, 1 glas cial of LIghtning Link Rapid mix
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 100 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.