missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Rhodamine Antibody Labeling Kit
Shop All Bio Techne ProductsDescription
- No desalting or dialysis steps
- Lightning-link Rapid technology can be used to label small quantities of antibody with 100% recovery
- Rhodamine used in kit has absorption peaks at 520 and 544nm
- Maximum emission is obtained when the rhodamine is excited at 544nm
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 300 μg |
| Product Type | Antibody Labeling Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?