missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) R-PE Antibody Labeling Kit

Product Code. 18747973 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg Reaction
1 x 100 μg Reaction
1 x 5 mg Reaction
3 x 0.01 mg Reaction
3 x 60 μg Reaction
5 x 1 mg Reaction
Unit Size:
1 set
3mg
600µg
60µg
This item is not returnable. View return policy

Product Code. 18747973

Brand: Novus Biologicals™ 7030010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Flow Cytometry

  • Easy to use
  • Only requires 30 seconds of hands-on time
  • No spin or separation steps
  • Generates conjugates identical to or better than those prepared with laborious multistep conjugation procedures
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 3 x 60 μg Reaction
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.