missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) PerCP Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
Unit Size:
1 set
100µg
1mg
Description
Peridinin-chlorophyll-protein complex (PerCP) is a small 35kDa phycobiliprotein isolated from red algae. It has a strong absorption peak at 482nm and a secondary absorption peak at 442nm. PerCP has a large stokes shift (195nm) with its maximum emission peak at 677nm. The protein is commonly used for fluorescent immunolabeling, especially in applications involving fluorescent-activated cell sorting.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 100 μg |
| Conjugate | PerCP |
| Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher reagent |
| Product Type | PerCP Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction