missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) PerCP Antibody Labeling Kit

Product Code. 18089673 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
Unit Size:
1 set
100µg
1mg
This item is not returnable. View return policy

Product Code. 18089673

Brand: Novus Biologicals™ 7180030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Flow Cytometry

Peridinin-chlorophyll-protein complex (PerCP) is a small 35kDa phycobiliprotein isolated from red algae. It has a strong absorption peak at 482nm and a secondary absorption peak at 442nm. PerCP has a large stokes shift (195nm) with its maximum emission peak at 677nm. The protein is commonly used for fluorescent immunolabeling, especially in applications involving fluorescent-activated cell sorting.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 30 μg
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher reagent
Product Type PerCP Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.