missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) PE-Texas Red Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
180 μg
30 μg
60 μg
600 μg
Unit Size:
1 set
600µg
60µg
Description
Lightning-Link is the easiest and quickest method available for making antibody (protein) conjugates, requiring just 30 seconds hands-on time. The antibody is covalently bonded to the label in a directional and controlled process at near-neutral pH.
PE/TEXAS RED is a tandem conjugate. The PE is excited at 488nm and functions as an energy donor for the TEXAS RED. Energy is transferred from the PE to the TEXAS RED via energy resonance transfer. The TEXAS RED emits the energy received from the PE in the form of long wavelength light at 615nm.
- The Lightning-Link conjugation kit allows PE-Texas Red tandem conjugations to set up in seconds, simply by adding a solution of the antibody to be labeled to a proprietary lyophilised mixture containing PE-Texas Red.
- The amount of antibody used for labeling should be the same as the pack size of LL-PE-Texas Red. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile.
- If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.
PE/TEXAS RED is a tandem conjugate. The PE is excited at 488nm and functions as an energy donor for the TEXAS RED. Energy is transferred from the PE to the TEXAS RED via energy resonance transfer. The TEXAS RED emits the energy received from the PE in the form of long wavelength light at 615nm.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 600 μg |
| Conjugate | PE/Texas Red |
| Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent |
| Product Type | PE-Texas Red Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction