missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) B-Phycoerythrin Antibody Labeling Kit

Product Code. 18758233 Shop All Bio Techne Products
Click to view available options
Quantity:
180 μg
60 μg
600 μg
Unit Size:
1 set
600µg
60µg
This item is not returnable. View return policy

Product Code. 18758233

Brand: Novus Biologicals™ 7160010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Flow Cytometry

B-Phycoerythrin (B-PE) is a fluorescent protein from phycobiliprotein family. It has a primary absorption peak at 546 nm and a secondary peak at 563 nm. When excited at 546 nm it has a strong fluorescence emission with its maximum at 575 nm.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 180 μg
Conjugate B-Phycoerythrin
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-B-PE Modifier reagent, 1 vial of LL-B-PE Quencher reagent
Product Type B-Phycoerythrin Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.