missing translation for 'onlineSavingsMsg'
Learn More

Amisulpride, MedChemExpress

Product Code. 16461126
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16461126

Brand: Medchem Express HY14545

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Amisulpride is a dopamine D2/D3 receptor antagonist with Kis of 2.8 and 3.2 nM for human dopamine D2 and D3, respectively.

Amisulpride is a dopamine D2/D3 receptor antagonist with Kis of 2.8 and 3.2 nM for human dopamine D2 and D3, respectively.

Specifications

Chemical Name or Material Amisulpride
CAS 71675-85-9
Color White
Purity Grade Notes Research
Molecular Formula C17H27N3O4S
Quantity 200 mg
Synonym DAN 2163
Solubility Information DMSO : 50 mg/mL (135.33 mM; Need ultrasonic) ∣H2O : 0.2 mg/mL (0.54 mM; Need ultrasonic)
SMILES CCN1C(CNC(C2=CC(S(=O)(CC)=O)=C(N)C=C2OC)=O)CCC1
Molecular Weight (g/mol) 369.48
Formula Weight 369.48
Percent Purity 99.56%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.