missing translation for 'onlineSavingsMsg'
Learn More

Human LIGHT/TNFSF14 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 4 publications

£184.00 - £543.00

Specifications

Antigen LIGHT/TNFSF14
Clone 115520
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Flow Cytometry, CyTOF, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18762959
GREENER_CHOICE
View Documents
R&D Systems
MAB664-SP
25 μg
£184.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18109190
GREENER_CHOICE
View Documents
R&D Systems
MAB664
500 μg
£543.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LIGHT/TNFSF14 Monoclonal specifically detects LIGHT/TNFSF14 in Human samples. It is validated for Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

LIGHT/TNFSF14
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TNFSF14, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
TNFSF14
IgG1
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
115520
Flow Cytometry, CyTOF, ELISA
Unconjugated
Mouse
Human
O43557
8740
Mouse myeloma cell line NS0-derived recombinant human LIGHT/TNFSF14 Asp74-Val240 Accession # O43557
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human LIGHT/TNFSF14 in ELISAs. In sandwich immunoassays, no significant cross-reactivity or interference with recombinant human (rh) Fas Ligand, rhAPRIL, recombinant mouse TRANCE, rhTNF-alpha or rhTRAIL is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.