Learn More
BINDER™ Light Module for Plants
Product Code. 30238310 Shop All BINDER Products
Change view
Click to view available options
Unit Size:
Each
Product Code. unitSize
30238310 Each
1 options
Product Code. 30238310 Supplier BINDER™ Supplier No. 80122439
  • ONLINE EXCLUSIVE PRICE
    £4965.00
    £4872.00 / Pack of 1
    Save £93.00
    1% Off
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
LED plant light module consisting of 16 LED strip lights
LED plant light module with 16 LED strip lights
Includes:
  • 3-channel controller
  • Fresh air pump
  • Independent power supply
Specifications
For Use With (Equipment) KB PRO 260, KB PRO 470, KB PRO 720, KB PRO 1060, KB PRO 1600, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600
Material Glass
Type Lighting
Features 16 LED Strip Lights