missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ 2D Light Module for Gbox XX6 and XX9
GREENER_CHOICE

Product Code. 12867004 Shop All Syngene Products
Change view
Click to view available options
For Use With (Equipment):
Gbox Chemi XX6/9
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12867004 Gbox Chemi XX6/9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12867004 Supplier Syngene™ Supplier No. DYVLIGHTMOD
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Syngene™ 2D Light Module for Gbox XX6 and XX9 is a CY dye edge light module(28x21cm) with low fluorescent plates for capture of 2D DIGE gels.

  • Required for multiplexing CY dyes
  • Contains Red, Green and Blue multiplex lights

Specifications

For Use With (Application) 2d DIGE electrophoresis imaging
For Use With (Equipment) Gbox Chemi XX6/9
Light Source LED
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.