missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Light Green SF, Yellowish, pure, high purity biological stain

Product Code. 10791501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10791501 10 g Glass bottle 10g
10269203 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10791501 Leverantör Thermo Scientific Chemicals Leverantörsnummer 229770100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5141-20-8 | C37H34N2Na2O9S3 | 792.844 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5141-20-8
Molecular Formula C37H34N2Na2O9S3
Molecular Weight (g/mol) 792.844
MDL Number MFCD00012121
InChI Key DGOBMKYRQHEFGQ-UHFFFAOYSA-L
Synonym Acid Green 5, C.I. 42095
PubChem CID 21223
ChEBI CHEBI:87065
IUPAC Name disodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-(4-sulfonatophenyl)methyl]anilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=C(C=C5)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 288.0°C
Absorption Ratio 0.96 to 1.16
Beilstein 14, 856
Merck Index 15, 5538
Solubility Information Solubility in water: 200g/L water (20°C). Other solubilities: soluble in alcohol
Infrared Spectrum Authentic
Lambda Max 629 to 634nm (H2O, 5mg/L, 1cm)
Formula Weight 792.84
Loss on Drying 10% max. (1 g, 110°C, 2 hrs)
Percent Purity ≥60% (TiCl3-titr.) (on substance as is)
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Purple
Quantity 10 g
Physical Form Powder, Crystals or Crystalline Powder
Chemical Name or Material Light Green SF, Yellowish, Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.