missing translation for 'onlineSavingsMsg'
Learn More

Manutan Light duty anti-fatigue ribbed pattern mat, grey 600mm x 910mm

Product Code. 12817459
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W):
600 x 910 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W)
12817459 600 x 910 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12817459 Supplier Manutan Supplier No. A353539

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
9. 4mm thickness anti-fatigue ribbed pattern for enhanced traction. Microcellular vinyl consists of millions of air bubbles providing basic worker comfort. All four sides are bevelled on mats (two sides on linear) and sealed to minimise tripping hazards (slip resistance R10 according to DIN 51130 and BG-rule BGR181). Free of toxic DOP with fire rating Class 2 according to NFPA Safety Code 101.
  • Grey
  • Weight: 2.3kg/m2
  • Mat thickness: 9.4mm
  • Dimensions (L x W): 600mm x 910mm

Specifications

Dimensions (L x W) 600 x 910 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.