missing translation for 'onlineSavingsMsg'
Learn More

Manutan Light duty anti-fatigue ribbed pattern mat, grey, 1200mm wide

Product Code. 12887539
Encompass_Preferred
Click to view available options
Width (Metric):
1200 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12887539

Brand: Manutan A353542

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
9. 4mm thickness anti-fatigue ribbed pattern for enhanced traction. Microcellular vinyl consists of millions of air bubbles providing basic worker comfort. All four sides are bevelled on mats (two sides on linear) and sealed to minimise tripping hazards (slip resistance R10 according to DIN 51130 and BG-rule BGR181). Free of toxic DOP with fire rating Class 2 according to NFPA Safety Code 101.
  • Grey
  • Weight: 2.3kg/m2
  • Mat thickness: 9.4mm
  • Width: 1200mm

Specifications

Width (Metric) 1200 mm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.