missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Light Cutter
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
- Accessory for BradyPrinter™ model BP-PR Plus series.
Specifications
Specifications
| Product Type | Cutter Tool |
| For Use With (Equipment) | BradyPrinter PR Plus Series Label Printer, THT 76 mm Core Printer |
| Description | Cutter CU4, field instlble |
| Format | Cutter |
| Quantity | 1 Ea. |
| Alternate Part Number | PR+-AUTOCUTTER-FI |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction