missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EVOS™ Light Cube, Cy™5.5
Click to view available options
:
Cy 5.5; Excitation/Emission: 4675/46nm, 794/16nm
Description
- Contains unique, proprietary LED light cubes
- Image fluorescent samples for specific wavelengthLight engine outputs high intensity over a short light path that delivers superior fluorophore excitation
- Each cube contains an LED, condensing optics, and hard-coated filters
- User-changeable and automatically recognized by system
- Shorter light path that affords superior detection of fluorescent signals
- Continuous illumination that enables consistent results
- 50,000+ hour bulb lifetime that affords lower operating costs
- Adjustable light intensity that helps reduce photobleaching
Specifications
Specifications
| Type | Light Cube |
| Product Line | EVOS |
| For Use With (Equipment) | Countess™ II FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System |
| Includes | Condensing Optics, Hard-Coated Filters, LED |
| Wavelength | Ex: 655/46 Em: 794/160 |
| Light Source | Metal Halide (Bulb) |
| Quantity | 1 each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
