Learn More
Invitrogen™ EVOS™ Light Cube, CFP-YFP EM
Product Code. 11907305
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
Product Code. Quantity unitSize
11907305 Each Each
1 options
Product Code. 11907305 Supplier Invitrogen™ Supplier No. AMEP4669
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
  • Contains unique, proprietary LED light cubes
  • Image fluorescent samples for specific wavelengthLight engine outputs high intensity over a short light path that delivers superior fluorophore excitation
  • Each cube contains an LED, condensing optics, and hard-coated filters
  • User-changeable and automatically recognized by system
  • Shorter light path that affords superior detection of fluorescent signals
  • Continuous illumination that enables consistent results
  • 50,000+ hour bulb lifetime that affords lower operating costs
  • Adjustable light intensity that helps reduce photobleaching
TRUSTED_SUSTAINABILITY
Specifications
Type Light Cube
Product Line EVOS
For Use With (Equipment) EVOS™ FL Color Imaging System, Countess™ II FL Automated Cell Counter, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Includes Condensing Optics, Hard-Coated Filters, LED
Wavelength Ex: 445/45 Em: 542/27
Light Source Metal Halide (Bulb)
Quantity Each