missing translation for 'onlineSavingsMsg'
Learn More

Leica Microsystems Light Bulb

Product Code. 11954978
Change view
Click to view available options
Wattage:
20 W
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wattage Unit Size
11954978 20 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11954978 Supplier Leica Microsystems Supplier No. 10447056

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Leica™ Light Bulbs are designed for use with Leica L2 Microscopes.

  • Halogen lamp 8 V/20 W
  • For Leica L2 Microscopes
TRUSTED_SUSTAINABILITY

Specifications

Voltage 8 V
Wattage 20 W
Product Type Light Bulb
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.