missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Light Attenuation Set For LB 9515

Product Code. 31777351 Shop All Berthold Technologies Products
Change view
Click to view available options
Color:
Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31777351 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31777351 Supplier Berthold Technologies Supplier No. 52401
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to calibrate NightSHADE and NightOWL imaging systems with a luminescence test plate.

  • For NightSHADE and NightOWL imaging systems.

Specifications

Color Gray
For Use With (Equipment) NightSHADE evo LB 985 in vivo Imaging System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.