missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIG1 Mouse, anti-Human, Unconjugated, Clone: 3D9G8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | LIG1 |
|---|---|
| Clone | 3D9G8 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17291496
|
Proteintech
67840-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17281496
|
Proteintech
67840-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIG1, also named as LRIG1, acts as a feedback negative regulator of signaling by receptor tyrosine kinases, through a mechanism that involves enhancement of receptor ubiquitination and accelerated intracellular degradation.
Specifications
| LIG1 | |
| 500 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| Primary |
| 3D9G8 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| 3978 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title