Learn More
JULABO™ Lift-Up Bath Cover for PURA™ Water Baths
Product Code. 16296611
Click to view available options
For Use With (Equipment):
PURA™ 10
PURA™ 14
PURA™ 22
PURA™ 30
PURA™ 4
Unit Size:
Each
Product Code. 16296611
Brand: JULABO™ 9970583
Only null left
Add to basket
Add to basket
Lift-up, transparent bath cover for PURA™ Water Bath up to 99.9°C. Protects against water loss through evaporation. The bath cover is made from plexiglass and polycarbonate.
  • Accessory for PURA™ Water Bath
  • Prevent water loss through evaporation
  • Protect samples from contamination
  • Made from plexiglass and polycarbonate
Specifications
Description Lift-up bath cover
For Use With (Equipment) PURA™ 22
Material Plexiglass and polycarbonate