Learn More
LIFR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22779-1-AP-20UL
 View more versions of this product
Product Code. 16802085
  • £135.00 / 20µL
Description
Signal-transducing molecule. May have a common pathway with IL6ST. The soluble form inhibits the biological activity of LIF by blocking its binding to receptors on target cells.
Specifications
Specifications
LIFR
Polyclonal
Unconjugated
LIFR
CD118, LIF R, LIF receptor, LIFR, SJS2, STWS, SWS
Rabbit
Antigen Affinity Chromatography
RUO
16880, 3977
-20°C
Liquid
Immunofluorescence, Immunoprecipitation, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P42702, P42703
LIFR
LIFR Fusion Protein Ag18742
20 μL
Primary
Human, Mouse