missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIFR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115511
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total LIFR.
Signal-transducing molecule. May have a common pathway with IL6ST. The soluble form inhibits the biological activity of LIF by blocking its binding to receptors on target cells.
Specifications
| LIFR | |
| Polyclonal | |
| Unconjugated | |
| Lifr | |
| A230075M04Rik; AW061234; CD118; CD118 antigen; D-factor/LIF receptor; FLJ98106; FLJ99923; leukemia inhibitor factor receptor alpha-chain; leukemia inhibitory factor receptor; leukemia inhibitory factor receptor alpha; leukemia inhibitory factor receptor alpha chain; LIF; LIF R beta; LIF receptor; Lifr; LIF-R; SJS2; soluble differentiation-stimulating factor receptor; STWS; SWS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16880, 3977, 81680 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O70535, P42702, P42703 | |
| Lifr | |
| A synthesized peptide derived from human LIFR(Accession P42702), corresponding to amino acid residues E319-Y369. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction