missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIFR Monoclonal Antibody (8E5E4D3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA529392
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human LIFR/CD118.
Signal-transducing molecule. May have a common pathway with IL6ST. The soluble form inhibits the biological activity of LIF by blocking its binding to receptors on target cells.
Specifications
| LIFR | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P42702 | |
| Lifr | |
| Recombinant Human LIFR protein (Met1-Ser833). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin) | |
| 8E5E4D3 | |
| Unconjugated | |
| Lifr | |
| A230075M04Rik; AW061234; CD118; CD118 antigen; D-factor/LIF receptor; FLJ98106; FLJ99923; leukemia inhibitor factor receptor alpha-chain; leukemia inhibitory factor receptor; leukemia inhibitory factor receptor alpha; leukemia inhibitory factor receptor alpha chain; LIF; LIF R beta; LIF receptor; Lifr; LIF-R; SJS2; soluble differentiation-stimulating factor receptor; STWS; SWS | |
| Mouse | |
| Protein A | |
| RUO | |
| 3977 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction