missing translation for 'onlineSavingsMsg'
Learn More

Lificiguat, MedChemExpress

Product Code. 16456258
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16456258

Brand: Medchem Express HY14927

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lificiguat binds to the β subunit of soluble guanylyl cyclase(sGC) with Kd of 0.6-1.1 μM in the presence of CO.

Lificiguat binds to the β subunit of soluble guanylyl cyclase(sGC) with Kd of 0.6-1.1 μM in the presence of CO.

Specifications

Chemical Name or Material Lificiguat
Physical Form Solid
CAS 170632-47-0
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H16N2O2
For Use With (Application) Cancer-Kinase/protease
Synonym YC-1
Solubility Information DMSO : ≥ 100 mg/mL (328.58 mM)
SMILES OCC1=CC=C(C2=NN(CC3=CC=CC=C3)C4=C2C=CC=C4)O1
Molecular Weight (g/mol) 304.34
Formula Weight 304.34
Percent Purity 98.82%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.