Learn More
Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, 300 to 710 μm
Lifetech™ ECR resins are robust polymeric resins designed for research and industrial immobilization of enzymes. Available for covalent, ionic, adsorption and affinity immobilization.
Brand:  Purolite™ LS01415-192
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 17049383
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Enzyme immobilization by adsorption, covalent, IEX and affinity
  • High mechanical stability
  • High porosity
  • Easily dried after immobilization
  • Suitable for batch and column configuration
  • Optimal for use in organic solvent/water free systems
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free
Specifications
Specifications
White to off-white spherical beads
400 to 650 Å
300 to 710 μm
Octadecyl
Lifetech™
Methacrylate (ECR Resin)
1 to 14
2°C to 20°C
57 to 68%
Wet
250 g