missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, 150 to 300 μm
Click to view available options
Quantity:
250 g
1 kg
Pore Size:
1200 to 1800 Å
300 to 600 Å
600 to 1200 Å
Unit Size:
1kg
250g
Description
- Enzyme immobilization by adsorption, covalent, IEX and affinity
- High mechanical stability
- High porosity
- Easily dried after immobilization
- Suitable for batch and column configuration
- Optimal for use in organic solvent/water free systems
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifications
Specifications
| pH Range | 3 to 10 |
| Pore Size | 1200 to 1800 Å |
| Content And Storage | 2°C to 20°C |
| Particle Size | 150 to 300 μm |
| Moisture | 70 to 80% |
| Functionality/Interaction Type | Amino |
| Form | Wet |
| Product Line | Lifetech |
| Quantity | 250 g |
| Type | Methacrylate (ECR Resin) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur